Thermo Scientific Chemicals

Thiazole-2-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H63329.03
1 g, Each
Thermo Scientific Chemicals

Thiazole-2-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H63329.03
1 g, Each
Quantity
Catalog number: H63329.03
also known as H63329-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10200-59-6
IUPAC Name
1,3-thiazole-2-carbaldehyde
Molecular Formula
C4H3NOS
InChI Key
ZGTFNNUASMWGTM-UHFFFAOYSA-N
SMILES
O=CC1=NC=CS1
Assay (GC)
>94.0%

Description

Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.

Solubility
Fully miscible with water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text