Chemical Identifiers
CAS133174-15-9
IUPAC Name5-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Molecular FormulaC21H23N3O5
InChI KeyNBMSMZSRTIOFOK-UHFFFAOYNA-N
SMILESNC(=O)NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more