3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(2-Naphthyl)-L-alanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in 1mol/L NaOH.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
S Hanzawa.; S Oe.; K Tokuhisa.; K Kawano. Chemo-enzymatic synthesis of 3-(2-naphthyl)-L-alanine by an aminotransferase from the extreme thermophile, Thermococcus profundus. Biotechnology Letters. 200123 (8), 589-591.
JE Rivier.; J Porter.; CL Rivier.; M Perrin. New effective gonadotropin releasing hormone antagonists with minimal potency for histamine release in vitro. J. Med. Chem. 198629 (10), 1846-1851.