Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
S Kobayashi.; J Nakano.; R Matsubara. Catalytic Mannich-type reaction of phosphorylimidates. Tetrahedron. 200966 (33), 6457-6461.
H Uchida.; A Chinone.; M Ohta. Reaction of 2-BenzoyIamino-2-methylpropionamidine and 2-Benzoylaminoacetamidine with Bifunctional Compounds. Bulletin of the Chemical Society of Japan. 197447 (7), 1720-1723.