4-Cyanoindole is used as a synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanoindole is used as a synthetic intermediate.
Solubility
Soluble in water.
Notes
Heat, light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
RUO – Research Use Only
C Wentrup. Hetarylnitrenes—IV: Annelated nitrenoazines. Tetrahedron,. 197127 (2), 367-374.
GS Ponticello.; JJ Baldwin. Useful synthesis of 4-substituted indoles. The Journal of Organic Chemistry. 197944 (22), 4003-4005.