L-Alanine 4-nitroanilide hydrochloride, 98%, Thermo Scientific Chemicals
L-Alanine 4-nitroanilide hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Alanine 4-nitroanilide hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number H63393.03
also known as H63393-03
Price (USD)/ Each
98.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
98.80
Add to cart
L-Alanine 4-nitroanilide hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH63393.03
Price (USD)/ Each
98.80
-
Add to cart
Chemical Identifiers
CAS31796-55-1
IUPAC Namehydrogen 2-amino-N-(4-nitrophenyl)propanamide chloride
Molecular FormulaC9H12ClN3O3
InChI KeyYEXRLSXNWLNHQR-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(N)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only