N-Boc-O-benzyl-D-tyrosine, 95%, Thermo Scientific Chemicals
N-Boc-O-benzyl-D-tyrosine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-O-benzyl-D-tyrosine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H63427.14
also known as H63427-14
Price (USD)/ Each
424.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
424.00
Add to cart
N-Boc-O-benzyl-D-tyrosine, 95%, Thermo Scientific Chemicals
Catalog numberH63427.14
Price (USD)/ Each
424.00
-
Add to cart
Chemical Identifiers
CAS63769-58-4
IUPAC Name(2S)-3-[4-(benzyloxy)phenyl]-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Molecular FormulaC21H25NO5
InChI KeyZAVSPTOJKOFMTA-LGWFVXIRNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only