N-Benzoyl-L-tyrosine ethyl ester, 98%, Thermo Scientific Chemicals
N-Benzoyl-L-tyrosine ethyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzoyl-L-tyrosine ethyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63434.06
also known as H63434-06
Price (USD)/ Each
35.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
35.40
Add to cart
N-Benzoyl-L-tyrosine ethyl ester, 98%, Thermo Scientific Chemicals
Catalog numberH63434.06
Price (USD)/ Each
35.40
-
Add to cart
Chemical Identifiers
CAS3483-82-7
IUPAC Nameethyl (2S)-3-(4-hydroxyphenyl)-2-(phenylformamido)propanoate
Molecular FormulaC18H19NO4
InChI KeySRLROPAFMUDDRC-XISACWJONA-N
SMILESCCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only