N-Fmoc-D-allo-isoleucine, 98%, Thermo Scientific Chemicals
N-Fmoc-D-allo-isoleucine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-D-allo-isoleucine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63455.03
also known as H63455-03
Price (USD)/ Each
198.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
198.00
Add to cart
N-Fmoc-D-allo-isoleucine, 98%, Thermo Scientific Chemicals
Catalog numberH63455.03
Price (USD)/ Each
198.00
-
Add to cart
Chemical Identifiers
CAS118904-37-3
IUPAC Name(2R,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoic acid
Molecular FormulaC21H23NO4
InChI KeyQXVFEIPAZSXRGM-KINVLMLONA-N
SMILESCC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate at -20°C. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Pilar E. Lópeza; Albert Isidro-Llobet; Carolina Gracia; Luis J. Cruz; Andrés García-Granados; Andrés Parra; Mercedes Álvarez; Fernando Albericio. Use of p-nitrobenzyloxycarbonyl (pNZ) as a permanent protecting group in the synthesis of Kahalalide F analogs. Tetrahedron Letters. 2005, 45(46), 7737-7741.
  2. Trent Conroy; Jin T. Guo; Prof. Roger G. Linington; Prof. Nicholas H. Hunt; and Dr. Richard J. Payne. Total Synthesis, Stereochemical Assignment, and Antimalarial Activity of Gallinamide A. Chemistry - A European Journal. 2011, 17(48), 13544-13552.