7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H63456.06
also known as H63456-06
Price (USD)/ Each
712.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
712.00
Add to cart
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH63456.06
Price (USD)/ Each
712.00
-
Add to cart
Chemical Identifiers
CAS73075-45-3
IUPAC Name7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Molecular FormulaC9H11Cl2N
InChI KeyOGIAIXMUSSACDB-UHFFFAOYSA-N
SMILESCl.ClC1=CC2=C(CCNC2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98%
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only