D-Glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals
D-Glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H63463.06
also known as H63463-06
Price (USD)/ Each
106.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
125.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
106.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
125.00 
Save 18.35 (15%)
Add to cart
D-Glutamic acid 5-tert-butyl ester, 95%, Thermo Scientific Chemicals
Catalog numberH63463.06
Price (USD)/ Each
106.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
125.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS45125-00-6
IUPAC Name2-amino-5-(tert-butoxy)-5-oxopentanoic acid
Molecular FormulaC9H17NO4
InChI KeyOIOAKXPMBIZAHL-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)CCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is employed as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. T J Dougherty; J A Thanassi; and M J Pucci. The Escherichia coli mutant requiring D-glutamic acid is the result of mutations in two distinct genetic loci. European J. Bacteriol. 1993, 175(1), 111-116.
  2. J. W. Goodman; and Danute E. Nitecki. Studies on the relation of a prior immune response to immunogenicity. Immunology. 1967, 13(6), 577-583.