Methyl thiazole-2-carboxylate, 95%, Thermo Scientific Chemicals
Methyl thiazole-2-carboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl thiazole-2-carboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63496.03
also known as H63496-03
Price (USD)/ Each
85.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
85.50
Add to cart
Methyl thiazole-2-carboxylate, 95%, Thermo Scientific Chemicals
Catalog numberH63496.03
Price (USD)/ Each
85.50
-
Add to cart
Chemical Identifiers
CAS55842-56-3
IUPAC Namemethyl 1,3-thiazole-2-carboxylate
Molecular FormulaC5H5NO2S
InChI KeyYRMZUOPMHMPNPG-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Suppliers CofA≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only