Thermo Scientific Chemicals

(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals

Catalog number: H63516.03
1 g, Each
Thermo Scientific Chemicals

(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid, 95%, Thermo Scientific Chemicals

Catalog number: H63516.03
1 g, Each
Quantity
Catalog number: H63516.03
also known as H63516-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
188970-92-5
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-{[(prop-2-en-1-yloxy)carbonyl]amino}propanoic acid
Molecular Formula
C22H22N2O6
InChI Key
MPVGCCAXXFLGIU-UHFFFAOYNA-N
SMILES
OC(=O)C(CNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (HPLC)
>94.0%
Appearance (Color)
White to off-white
Form
Powder

Description

(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text