Thermo Scientific Chemicals

Methyl 2-bromothiazole-4-carboxylate, 96%, Thermo Scientific Chemicals

Catalog number: H63541.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-bromothiazole-4-carboxylate, 96%, Thermo Scientific Chemicals

Catalog number: H63541.06
5 g, Each
Quantity
Catalog number: H63541.06
also known as H63541-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
170235-26-4
IUPAC Name
methyl 2-bromo-1,3-thiazole-4-carboxylate
Molecular Formula
C5H4BrNO2S
InChI Key
YOWKNNKTQWCYNC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CSC(Br)=N1
Assay (HPLC)
>95.0%

Description

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text