Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
H. R. Snyder; E. P. Merica; C. G. Force; E. G. White. Synthesis of Indoles by Catalytic Reduction of o-Nitrobenzyl Cyanides. J. Am. Chem. Soc. 1958, 80 (17), 4622-4625.