4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals
4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H63572.MD
also known as H63572-MD
Price (USD)/ Each
53.30
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
53.30
Add to cart
4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals
Catalog numberH63572.MD
Price (USD)/ Each
53.30
-
Add to cart
Chemical Identifiers
CAS167479-78-9
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White to off-white
Assay by NMR>94.0%
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Christopher G. Bazewicz; Jacob S. Lipkin; Emily E. Smith; Melanie T. Liskov; and Scott H. Brewer. Expanding the Utility of 4-Cyano-l-Phenylalanine As a Vibrational Reporter of Protein Environments. J. Phys. Chem.B. 2012, 116 (35), 10824-10831.
  2. Nicole Maurici; Andrew Dippel; Melanie Liskov; Scott Brewer; Christine Phillips-Piro. Probing Structural Implications of Unnatural Amino Acid Incorporation into Green Fluorescent Protein. Bio physical journal. 2015, 108 (2), 511a.