2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Xing-Xiu Chen; Feng Gao; Qi Wang; Xing Huang; Dan Wang. Design, synthesis and biological evaluation of paclitaxel-mimics possessing only the oxetane D-ring and side chain structures. Fitoterapia. 1997, 92 111-115.
Michael Gurry; Patrick McArdle; and Fawaz Aldabbagh. Synthesis of a Spirocyclic Oxetane-Fused Benzimidazole. Molecules. 2015 2013864-13874.