Thermo Scientific Chemicals

2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%, Thermo Scientific Chemicals

Catalog number: H63578.06
5 g, Each
Thermo Scientific Chemicals

2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%, Thermo Scientific Chemicals

Catalog number: H63578.06
5 g, Each
Quantity
Catalog number: H63578.06
also known as H63578-06
Price (USD)
Quantity
-
Have Questions?

Description

2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text