L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals
L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63598.03
also known as H63598-03
Price (USD)/ Each
65.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
65.50
Add to cart
L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH63598.03
Price (USD)/ Each
65.50
-
Add to cart
Chemical Identifiers
CAS4561-10-8
IUPAC Namehydrogen 1,5-dibenzyl (2S)-2-aminopentanedioate chloride
Molecular FormulaC19H22ClNO4
InChI KeyGRGJVECUQLAEDM-LMOVPXPDSA-N
SMILES[H+].[Cl-].N[C@@H](CCC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>97.5%
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. R. M. Kondratenko; L. A. BaltinaJr; L. A. Baltina; N. Zh. Baschenko; G. A. Tolstikov. Synthesis and immunomodulating activity of new glycopeptides of glycyrrhizic acid containing residues of L-glutamic acid. Russian Journal of Bioorganic Chemistry. 2006, 32(6), 595-601.
  2. James E. Shields; William H. Mcgregor; Frederick H. Carpenter. Preparation of Benzyl and p-Nitrobenzyl Esters of Amino Acids. J. Org. Chem. 1961, 26(5), 1491-1494.