Thermo Scientific Chemicals

L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63598.06
5 g, Each
Thermo Scientific Chemicals

L-Glutamic acid dibenzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63598.06
5 g, Each
Quantity
Catalog number: H63598.06
also known as H63598-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4561-10-8
IUPAC Name
hydrogen 1,5-dibenzyl (2S)-2-aminopentanedioate chloride
Molecular Formula
C19H22ClNO4
InChI Key
GRGJVECUQLAEDM-LMOVPXPDSA-N
SMILES
[H+].[Cl-].N[C@@H](CCC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Assay (Titration)
>97.5%

Description

L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text