Thermo Scientific Chemicals

Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%, Thermo Scientific Chemicals

Catalog number: H63609.06
5 g, Each
Thermo Scientific Chemicals

Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%, Thermo Scientific Chemicals

Catalog number: H63609.06
5 g, Each
Quantity
Catalog number: H63609.06
also known as H63609-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
146987-10-2
IUPAC Name
(2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular Formula
C31H38N4O6S
InChI Key
OFIBQNGDYNGUEZ-CVKYCUBCNA-N
SMILES
[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2
Assay (HPLC)
>94.0%

Description

Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

Solubility
Slightly soluble in water and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text