Thermo Scientific Chemicals

Thiazole-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H63620.06
5 g, Each
Thermo Scientific Chemicals

Thiazole-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H63620.06
5 g, Each
Quantity
Catalog number: H63620.06
also known as H63620-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14190-59-1
IUPAC Name
1,3-thiazole-2-carboxylic acid
Molecular Formula
C4H3NO2S
InChI Key
IJVLVRYLIMQVDD-UHFFFAOYSA-N
SMILES
OC(=O)C1=NC=CS1
Assay (HPLC)
>94.0%

Description

Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text