N-Boc-D-threonine, 95%, Thermo Scientific Chemicals
N-Boc-D-threonine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-D-threonine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63647.06
also known as H63647-06
Price (USD)/ Each
96.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
96.10
Add to cart
N-Boc-D-threonine, 95%, Thermo Scientific Chemicals
Catalog numberH63647.06
Price (USD)/ Each
96.10
-
Add to cart
Chemical Identifiers
CAS55674-67-4
IUPAC Name(2R,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoic acid
Molecular FormulaC9H17NO5
InChI KeyLLHOYOCAAURYRL-NTSWFWBYSA-N
SMILESC[C@H](O)[C@@H](NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is known to inhibit growth and cell wall synthesis of Mycobacterium smegmatis. Also used as a synthetic intermediate for the production of chiral antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is known to inhibit growth and cell wall synthesis of Mycobacterium smegmatis. Also used as a synthetic intermediate for the production of chiral antibiotics.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Kunihiko Yabu; and Shozo Takahashi. Inhibition of Growth and Cell Wall Synthesis of Mycobacterium smegmatis by D-Threonine. Microbiology and Immunology. 1978, 22(2), 103-107.
  2. V. F. Pozdnev; N. N. Podgornova; N. K. Zentsova; G. I. Aukone; U. O. Kalei. Preparation of tert-butoxycarbonyl derivatives of amino acids using di-tert-butyl pyrocarbonate. Chemistry of Natural Compounds. 1979, 15(4), 471-476.