Thermo Scientific Chemicals

3-(2-Aminoethyl)-5-bromoindole, 97%, Thermo Scientific Chemicals

Catalog number: H63653.03
1 g, Each
Thermo Scientific Chemicals

3-(2-Aminoethyl)-5-bromoindole, 97%, Thermo Scientific Chemicals

Catalog number: H63653.03
1 g, Each
Quantity
Catalog number: H63653.03
also known as H63653-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3610-42-2
IUPAC Name
2-(5-bromo-1H-indol-3-yl)ethan-1-amine
Molecular Formula
C10H11BrN2
InChI Key
CGHUQJRRADEHTQ-UHFFFAOYSA-N
SMILES
NCCC1=CNC2=CC=C(Br)C=C12
Form
Powder
Proton NMR
Conforms to structure
Appearance (Color)
White to yellow
Assay from Suppliers CofA
≥95.0%

Description

N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text