Thermo Scientific Chemicals

D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H63685.22
100 g, Each
Thermo Scientific Chemicals

D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H63685.22
100 g, Each
Quantity
Catalog number: H63685.22
also known as H63685-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5874-57-7
IUPAC Name
4-bromo-N-{1-[3-(4-bromophenyl)-4-oxo-3,4-dihydroquinazolin-2-yl]ethyl}-N-methylbenzene-1-sulfonamide
Molecular Formula
C23H19Br2N3O3S
InChI Key
LYRWWUCOZBTOPL-UHFFFAOYNA-N
SMILES
CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥94.0%
Optical Rotation
-4.0 to -6.0°
Form
Powder or crystals or crystalline powder
Melting Point
163.0-168.0°C

Description

D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text