L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.
Solubility
Soluble in methanol (50gmg/ml).
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
M Antoniou; C Poulos; T Tsegenidis. Synthesis and biological activity of analogues of the C-terminal hexapeptide of substance P with modifications at glutaminyl and methioninyl residues. Structure-activity studies. International Journal of Peptide and Protein Reseach. 1992, 40(5), 395-400.
A Manolopoulou; K Karagiannis; G Stavropoulos; C Poulos; C C Jordan; R M Hagan. Synthesis of potent antagonists of substance P by modifying the methionyl and glutaminyl residues of its C-terminal hexapeptide and without using D-amino acids. International Journal of Peptide and Protein Reseach. 1993, 41(4), 411-414.