Thermo Scientific Chemicals

L-Aspartic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63716.14
25 g, Each
Thermo Scientific Chemicals

L-Aspartic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63716.14
25 g, Each
Quantity
Catalog number: H63716.14
also known as H63716-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1791-13-5
IUPAC Name
1,4-di-tert-butyl 2-aminobutanedioate hydrochloride
Molecular Formula
C12H24ClNO4
InChI Key
GVLZIMQSYQDAHB-UHFFFAOYNA-N
SMILES
Cl.CC(C)(C)OC(=O)CC(N)C(=O)OC(C)(C)C
Assay (HPLC)
>97.5%

Description

L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid di-tert-butyl ester hydrochloride may be used in neurological comparison studies with structurally similar compounds such as L-Glutamic acid di-tert-butyl ester.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text