Thermo Scientific Chemicals

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%, Thermo Scientific Chemicals

Catalog number: H63726.06
5 g, Each
Thermo Scientific Chemicals

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%, Thermo Scientific Chemicals

Catalog number: H63726.06
5 g, Each
Quantity
Catalog number: H63726.06
also known as H63726-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
252049-08-4
IUPAC Name
4-{[(benzyloxy)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Molecular Formula
C27H28N2O5
InChI Key
KKUPBFMKUPMFRE-UHFFFAOYNA-N
SMILES
OC(=O)C(CCNCOCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Assay (unspecified)
≥94.0%

Description

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text