Thermo Scientific Chemicals

Nepsilon-Boc-L-lysine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63727.03
1 g, Each
Thermo Scientific Chemicals

Nepsilon-Boc-L-lysine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63727.03
1 g, Each
Quantity
Catalog number: H63727.03
also known as H63727-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2389-48-2
IUPAC Name
methyl (2S)-2-amino-6-{[(tert-butoxy)carbonyl]amino}hexanoate hydrochloride
Molecular Formula
C12H25ClN2O4
InChI Key
NANRHOPPXCBHGI-MVRPGTNWNA-N
SMILES
Cl.COC(=O)[C@@H](N)CCCCNC(=O)OC(C)(C)C
Assay (HPLC)
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in methanol.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text