2,3-Dichloropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
2,3-Dichloropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dichloropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H63733.06
also known as H63733-06
Price (USD)/ Each
318.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
318.00
Add to cart
2,3-Dichloropyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberH63733.06
Price (USD)/ Each
318.00
-
Add to cart
Chemical Identifiers
CAS184416-84-0
IUPAC Name2,3-dichloropyridine-4-carboxylic acid
Molecular FormulaC6H3Cl2NO2
InChI KeyVGKZZKKTZKRCPA-UHFFFAOYSA-N
SMILESOC(=O)C1=C(Cl)C(Cl)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only