Chemical Identifiers
CAS146982-27-6
IUPAC Name(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(prop-2-en-1-yloxy)carbonyl]amino}hexanoic acid
Molecular FormulaC25H28N2O6
InChI KeyOJBNDXHENJDCBA-ANBDAQEENA-N
SMILESOC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more