Chemical Identifiers
CAS13122-90-2
IUPAC Name2-(2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanamido)-3-phenylpropanoic acid
Molecular FormulaC23H28N2O5
InChI KeyNNOBHAOOLCEJBL-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(O)=O
View more