Thermo Scientific Chemicals

N-Boc-L-phenylalanyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H63788.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-L-phenylalanyl-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog number: H63788.06
5 g, Each
Quantity
Catalog number: H63788.06
also known as H63788-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13122-90-2
IUPAC Name
2-(2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanamido)-3-phenylpropanoic acid
Molecular Formula
C23H28N2O5
InChI Key
NNOBHAOOLCEJBL-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay from Suppliers CofA
≥94.0% (NMR)
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text