3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.
Solubility
Soluble in dilute aqeous acid and dilute aqeous base.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
RUO – Research Use Only
Menek Goldstein,; Zvi Weiss.Inhibition of tyrosine hydroxylase by 3-iodo-L-tyrosine. Life Sciences. publication19654(2) 261-264.
Kensaku Sakamoto,; Kazutaka Murayama,; Kenji Oki,; Fumie Iraha,; Miyuki Kato-Murayama,; Masahiro Takahashi,; Kazumasa Ohtake. Genetic Encoding of 3-Iodo-l-Tyrosine in Escherichia coli for Single-Wavelength Anomalous Dispersion Phasing in Protein Crystallography. Structure. 2009, 17 (3) ,335-344.