tert-Butyl 2-aminoisobutyrate hydrochloride, 98%, Thermo Scientific Chemicals
tert-Butyl 2-aminoisobutyrate hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl 2-aminoisobutyrate hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H63817.06
also known as H63817-06
Price (USD)/ Each
587.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
587.00
Add to cart
tert-Butyl 2-aminoisobutyrate hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH63817.06
Price (USD)/ Each
587.00
-
Add to cart
Chemical Identifiers
CAS84758-81-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only