Thermo Scientific Chemicals

Thiophene-2-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H63819.14
25 g, Each
Thermo Scientific Chemicals

Thiophene-2-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H63819.14
25 g, Each
Quantity
Catalog number: H63819.14
also known as H63819-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
193978-23-3
IUPAC Name
4,4,5,5-tetramethyl-2-(thiophen-2-yl)-1,3,2-dioxaborolane
Molecular Formula
C10H15BO2S
InChI Key
FFZHICFAHSDFKZ-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=CS1
Assay from Supplier's CofA
≥97.5%
Appearance (Color)
White to pale cream
Form
Powder or crystals or crystalline powder
Melting Point
64.0-71.0°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text