Thermo Scientific Chemicals
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%, Thermo Scientific Chemicals
Catalog number: H63825.MD
250 mg, Each
Thermo Scientific Chemicals
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%, Thermo Scientific Chemicals
Catalog number: H63825.MD
250 mg, Each
Quantity
Catalog number: H63825.MD
also known as H63825-MD
Price (USD)
Quantity
-
Have Questions?
Description
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.