Thermo Scientific Chemicals

Nalpha-Boc-L-lysine, 97%, Thermo Scientific Chemicals

Catalog number: H63859.22
100 g, Each
Thermo Scientific Chemicals

Nalpha-Boc-L-lysine, 97%, Thermo Scientific Chemicals

Catalog number: H63859.22
100 g, Each
Quantity
Catalog number: H63859.22
also known as H63859-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13734-28-6
IUPAC Name
(2S)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Molecular Formula
C11H22N2O4
InChI Key
DQUHYEDEGRNAFO-SVGMAFHSNA-N
SMILES
CC(C)(C)OC(=O)N[C@@H](CCCCN)C(O)=O
Assay (HPLC)
>96.0%

Description

N-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-lysine is a protected analogue of a major amino acid participating in the binding of AcH to proteins. N-Boc-L-lysine as well as other t-butoxycarbonyl (Boc) amino acids can be converted to their respective amino acids via cleavage of the urethane bond.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text