Thermo Scientific Chemicals

4-(4-Fluorobenzyl)piperidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H63860.03
1 g, Each
Thermo Scientific Chemicals

4-(4-Fluorobenzyl)piperidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H63860.03
1 g, Each
Quantity
Catalog number: H63860.03
also known as H63860-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
193357-52-7
Appearance (Color)
White
Form
Solid
Assay (HPLC)
>96.0%

Description

Reactant in the synthesis of 4-(Substituted-benzyl)piperidines and (±)-3-(Substituted-benzyl)pyrrolidines. It an important and valuable pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of 4-(Substituted-benzyl)piperidines and (±)-3-(Substituted-benzyl)pyrrolidines. It an important and valuable pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text