5-(Di-tert-butylphosphino)-1', 3', 5'-triphenyl-1'H-[1,4']bipyrazole is a non-proprietary ligand for palladium-catalyzed amination of aryl halides including aryl chlorides. Works best when the palladium source is Pd2(dba)3
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-(Di-tert-butylphosphino)-1′, 3′, 5′-triphenyl-1′H-[1,4′]bipyrazole is a non-proprietary ligand for palladium-catalyzed amination of aryl halides including aryl chlorides. Works best when the palladium source is Pd2(dba)3
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Saravanan Gowrisankar; Alexey G Sergeev; Pazhamalai Anbarasan; Anke Spannenberg; Helfried Neumann; Matthias Beller. A general and efficient catalyst for palladium-catalyzed C-O coupling reactions of aryl halides with primary alcohols. Journal of the American Chemical Society. 2010 132(33), 11592-11598.
Brian J Kotecki; Dilinie P Fernando; Anthony R Haight; Kirill A Lukinr. A general method for the synthesis of unsymmetrically substituted ureas via palladium-catalyzed amidation. Organic Letters. 2009. 11(4), 947-950.