Thermo Scientific Chemicals

2-Bromopyridine-5-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H63867.03
1 g, Each
Thermo Scientific Chemicals

2-Bromopyridine-5-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H63867.03
1 g, Each
Quantity
Catalog number: H63867.03
also known as H63867-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149806-06-4
Assay (HPLC)
>94.0%

Description

A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful synthetic intermediate. 6-Bromonicotinaldehyde is used in wittig reaction. Hydrodehalogenation of 6-bromonicotinaldehyde to produce alcohol.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. 2-7°C Refrigerate.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text