Thermo Scientific Chemicals

2-Bromo-4,5-difluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63902.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-4,5-difluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63902.06
5 g, Each
Quantity
Catalog number: H63902.06
also known as H63902-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
883502-07-6
Assay (HPLC)
>97.5%

Description

2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,5-difluorophenylacetic acid is used as intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text