O-Benzyl-L-tyrosine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
O-Benzyl-L-tyrosine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

O-Benzyl-L-tyrosine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H63908.06
also known as H63908-06
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
O-Benzyl-L-tyrosine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Catalog numberH63908.06
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS34805-17-9
IUPAC Namemethyl (2S)-2-amino-3-[4-(benzyloxy)phenyl]propanoate hydrochloride
Molecular FormulaC17H20ClNO3
InChI KeyIQKXGACPIPNLEL-RDZNSINMNA-N
SMILESCl.COC(=O)[C@@H](N)CC1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only