Chemical Identifiers
CAS6968-11-2
IUPAC Name2-{[(benzyloxy)carbonyl]amino}-3-[(2-{[(benzyloxy)carbonyl]amino}-2-carboxyethyl)disulfanyl]propanoic acid
Molecular FormulaC22H24N2O8S2
InChI KeyPTRQEEVKHMDMCF-UHFFFAOYNA-N
SMILESOC(=O)C(CSSCC(NC(=O)OCC1=CC=CC=C1)C(O)=O)NC(=O)OCC1=CC=CC=C1
View more