A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A chiral arylalkylamine used as organocatalysts. Used in the synthesis of chiral diamines also in determination of chiral recognition properties of novel fluorescent chemosensors for amino acid. an efficient and practical synthesis of l-. alpha.-amino acids using (r)-phenylglycinol as a chiral auxiliary.
Solubility
Soluble in chloroform, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
- Rituraj Dubey,; Ravi Bhushan. A Rapid, Robust and Ultra-Sensitive HPLC Enantioseparation of β-Amino Alcohols. J Chromatogr Sci 2015,
- Mercedes Amat Prof. Dr.,; Margalida Cantó Dr.,; Núria Llor Dr.,; Viviana Ponzo Dr.,; Maria Pérez,; Joan Bosch Prof. Dr. Dynamic Kinetic Resolution and Desymmetrization of Enantiotopic Groups by Cyclodehydration of Racemic or Prochiral δ-Oxoesters with (R)-Phenylglycinol: Enantioselective Synthesis of Piperidines. TOC. 2002, 114 (2), 345-348.