3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals
3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number H63941.03
also known as H63941-03
Price (USD)/ Each
177.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
177.00
Add to cart
3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals
Catalog numberH63941.03
Price (USD)/ Each
177.00
-
Add to cart
Chemical Identifiers
CAS64090-98-8
IUPAC Name(2S)-2-amino-3-(pyridin-3-yl)propanoic acid
Molecular FormulaC8H10N2O2
InChI KeyDFZVZEMNPGABKO-ZETCQYMHSA-N
SMILESN[C@@H](CC1=CC=CN=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Tingting Huang,; Yemin Wang ,; Yin,; Yanhua Du,; Meifeng Tao,; Jing Xu,; Wenqing Chen,; Shuangjun Lin,; Zixin Deng. Identification and Characterization of the Pyridomycin Biosynthetic Gene Cluster of Streptomyces pyridomyceticus NRRL B-2517title . The Journal of Biological Chemistry, 2011, 286, 20648-20657.
  2. Hu, Jialiang; Yan, Ming; Pu, Chunyan; Wang, Jingjing; E. Van den Steen, Philippe; Opdenakker, Ghislain; Xu, Hanmei. Chemically Synthesized Matrix Metalloproteinase and Angiogenesis-inhibiting Peptides as Anticancer Agents. / Anti-Cancer Agents in Medicinal Chemistry (Formerly Current Medicinal Chemistry - Anti-Cancer Agents. 2013, 14(3), 483-494.