Thermo Scientific Chemicals

3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals

Catalog number: H63941.MD
250 mg, Each
Thermo Scientific Chemicals

3-(3-Pyridyl)-L-alanine, 98%, Thermo Scientific Chemicals

Catalog number: H63941.MD
250 mg, Each
Quantity
Catalog number: H63941.MD
also known as H63941-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64090-98-8
IUPAC Name
(2S)-2-amino-3-(pyridin-3-yl)propanoic acid
Molecular Formula
C8H10N2O2
InChI Key
DFZVZEMNPGABKO-ZETCQYMHSA-N
SMILES
N[C@@H](CC1=CC=CN=C1)C(O)=O
Assay (HPLC)
>97.5%

Description

3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text