A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Ying-Ling Liu; Shr-Hau Tsai; Chuan-Shao Wu; and Ru-Jong Jeng. Preparation and characterization of hyperbranched polyaspartimides from bismaleimides and triamines. Journal of Polymer Science Part A: Polymer Chemistry. 2004 42 (23), 5921-5928.
Zhonggang Wang; Bufeng Zhang; Hao Yu; Guiyang Lia; and Yajie Bao. Synthetic control of network topology and pore structure in microporous polyimides based on triangular triphenylbenzene and triphenylamine units. Soft Matter. 2011 7 5723-5730.