Thermo Scientific Chemicals

Tris(4-aminophenyl)amine, 97%, Thermo Scientific Chemicals

Catalog number: H63963.MD
250 mg, Each
Thermo Scientific Chemicals

Tris(4-aminophenyl)amine, 97%, Thermo Scientific Chemicals

Catalog number: H63963.MD
250 mg, Each
Quantity
Catalog number: H63963.MD
also known as H63963-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5981-09-9
IUPAC Name
N1,N1-bis(4-aminophenyl)benzene-1,4-diamine
Molecular Formula
C18H18N4
InChI Key
SNLFYGIUTYKKOE-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)N(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
Assay (HPLC)
>96.0%

Description

A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A fluorescent trifunctional highly symmetric probe. Shows remarkably high nonlinear optical properties.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text