7-Bromoquinoline, 95%, Thermo Scientific Chemicals
7-Bromoquinoline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Bromoquinoline, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H63987.06
also known as H63987-06
Price (USD)/ Each
456.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
456.00
Add to cart
7-Bromoquinoline, 95%, Thermo Scientific Chemicals
Catalog numberH63987.06
Price (USD)/ Each
456.00
-
Add to cart
Chemical Identifiers
CAS4965-36-0
IUPAC Name7-bromoquinoline
Molecular FormulaC9H6BrN
InChI KeyXYBSZCUHOLWQQU-UHFFFAOYSA-N
SMILESBrC1=CC=C2C=CC=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only